Assay Details
Gene Symbol: ACTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031229.1 | NP_001026400.1 | 4 - 5 | 449 | 91 | |
XM_015288391.2 | XP_015143877.1 | 5 - 6 | 537 | 91 | ||
XM_015288392.2 | XP_015143878.1 | 5 - 6 | 520 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
