Assay Details
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031267.1 | NP_001026438.1 | 3 - 4 | 1918 | 69 | |
GenBank mRNA | AJ720086.1 | - | 3 - 4 | 1918 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
