Assay Details
Gene Symbol: AGRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031457.1 | NP_001026628.1 | 1 - 2 | 210 | 82 | |
XM_025154207.1 | XP_025009975.1 | 2 - 3 | 309 | 82 | ||
GenBank mRNA | AB489991.1 | - | 2 - 3 | 251 | 82 | |
AB489992.1 | - | 2 - 3 | 334 | 82 | ||
AB489993.1 | - | 1 - 2 | 668 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
