Assay Details
Gene Symbol: ELOVL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031539.1 | NP_001026710.1 | 2 - 3 | 359 | 113 | |
GenBank mRNA | AJ720366.1 | - | 2 - 3 | 359 | 113 | |
HQ264103.1 | - | 2 - 3 | 329 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
