Assay Details
Gene Symbol: FASLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031559.1 | NP_001026730.1 | 1 - 2 | 283 | 88 | |
GenBank mRNA | AJ890143.1 | - | 1 - 2 | 283 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
