Assay Details
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030700.1 | NP_001025871.1 | 4 - 5 | 658 | 79 | |
GenBank mRNA | AJ720296.1 | - | 4 - 5 | 658 | 79 | |
CR523234.1 | - | 3 - 4 | 350 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
