Assay Details
Gene Symbol: NAPEPLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030730.1 | NP_001025901.1 | 4 - 5 | 1292 | 102 | |
XM_015275728.2 | XP_015131214.1 | 4 - 5 | 1751 | 102 | ||
XM_015275729.2 | XP_015131215.1 | 4 - 5 | 1256 | 102 | ||
GenBank mRNA | AJ719820.1 | - | 4 - 5 | 1292 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
