Assay Details
Gene Symbol: NEDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030750.1 | NP_001025921.1 | 10 - 11 | 1516 | 90 | |
XM_015283802.2 | XP_015139288.1 | 10 - 11 | 1445 | 90 | ||
XM_015283806.2 | XP_015139292.1 | 10 - 11 | 1452 | 90 | ||
XM_015283818.2 | XP_015139304.1 | 10 - 11 | 1441 | 90 | ||
XM_015283822.2 | XP_015139308.1 | 10 - 11 | 1448 | 90 | ||
XM_025148212.1 | XP_025003980.1 | 10 - 11 | 1495 | 90 | ||
GenBank mRNA | AJ851532.1 | - | 10 - 11 | 1516 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
