Assay Details
Gene Symbol: MTSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030954.1 | NP_001026125.1 | 4 - 5 | 799 | 85 | |
XM_015283091.2 | XP_015138577.1 | 4 - 5 | 836 | 85 | ||
XM_015283092.2 | XP_015138578.1 | 4 - 5 | 836 | 85 | ||
XM_015283093.2 | XP_015138579.1 | 4 - 5 | 836 | 85 | ||
XM_015283094.2 | XP_015138580.1 | 4 - 5 | 836 | 85 | ||
XM_015283095.2 | XP_015138581.1 | 4 - 5 | 836 | 85 | ||
XM_015283097.2 | XP_015138583.1 | 4 - 5 | 836 | 85 | ||
XM_015283098.2 | XP_015138584.1 | 4 - 5 | 836 | 85 | ||
XM_015283099.2 | XP_015138585.1 | 4 - 5 | 836 | 85 | ||
XM_015283100.2 | XP_015138586.1 | 4 - 5 | 836 | 85 | ||
XM_015283101.2 | XP_015138587.1 | 4 - 5 | 836 | 85 | ||
XM_015283102.2 | XP_015138588.1 | 4 - 5 | 836 | 85 | ||
XM_015283103.2 | XP_015138589.1 | 4 - 5 | 836 | 85 | ||
XM_015283104.2 | XP_015138590.1 | 4 - 5 | 836 | 85 | ||
XM_015283105.2 | XP_015138591.1 | 4 - 5 | 836 | 85 | ||
XM_025147131.1 | XP_025002899.1 | 4 - 5 | 363 | 85 | ||
GenBank mRNA | AJ719272.1 | - | 4 - 5 | 799 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
