Assay Details
Gene Symbol: MTSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030954.1 | NP_001026125.1 | 6 - 7 | 969 | 150 | |
XM_015283094.2 | XP_015138580.1 | 6 - 7 | 1006 | 150 | ||
XM_015283099.2 | XP_015138585.1 | 6 - 7 | 1006 | 150 | ||
XM_015283103.2 | XP_015138589.1 | 6 - 7 | 1006 | 150 | ||
GenBank mRNA | AJ719272.1 | - | 6 - 7 | 969 | 150 | |
CR353915.1 | - | 2 - 3 | 146 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
