Assay Details
Gene Symbol: TNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031166.2 | NP_001026337.2 | 1 - 2 | 509 | 66 | |
XM_015285792.2 | XP_015141278.1 | 1 - 2 | 3090 | 66 | ||
GenBank mRNA | AJ851742.1 | - | 1 - 2 | 515 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
