Assay Details
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031281.1 | NP_001026452.1 | 3 - 4 | 435 | 94 | |
GenBank mRNA | AJ719913.1 | - | 3 - 4 | 435 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
