Assay Details
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031404.1 | NP_001026575.1 | 2 - 3 | 106 | 118 | |
XM_015277296.2 | XP_015132782.1 | 2 - 3 | 95 | 118 | ||
XM_015277303.2 | XP_015132789.1 | 2 - 3 | 95 | 118 | ||
GenBank mRNA | AJ851542.1 | - | 2 - 3 | 106 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
