Assay Details
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031404.1 | NP_001026575.1 | 3 - 4 | 254 | 146 | |
XM_015277296.2 | XP_015132782.1 | 3 - 4 | 243 | 146 | ||
XM_015277297.2 | XP_015132783.1 | 2 - 3 | 500 | 146 | ||
XM_015277298.2 | XP_015132784.1 | 3 - 4 | 372 | 146 | ||
XM_015277299.2 | XP_015132785.1 | 2 - 3 | 225 | 146 | ||
XM_015277301.2 | XP_015132787.1 | 2 - 3 | 194 | 146 | ||
XM_015277302.2 | XP_015132788.1 | 2 - 3 | 417 | 146 | ||
XM_015277303.2 | XP_015132789.1 | 3 - 4 | 243 | 146 | ||
GenBank mRNA | AJ851542.1 | - | 3 - 4 | 254 | 146 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015277301.2, XM_015277302.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
