Assay Details
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031404.1 | NP_001026575.1 | 4 - 5 | 352 | 74 | |
XM_015277296.2 | XP_015132782.1 | 4 - 5 | 341 | 74 | ||
XM_015277297.2 | XP_015132783.1 | 3 - 4 | 598 | 74 | ||
XM_015277298.2 | XP_015132784.1 | 4 - 5 | 470 | 74 | ||
XM_015277299.2 | XP_015132785.1 | 3 - 4 | 323 | 74 | ||
XM_015277301.2 | XP_015132787.1 | 3 - 4 | 292 | 74 | ||
XM_015277302.2 | XP_015132788.1 | 3 - 4 | 515 | 74 | ||
XM_015277303.2 | XP_015132789.1 | 4 - 5 | 341 | 74 | ||
GenBank mRNA | AJ851542.1 | - | 4 - 5 | 352 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
