Assay Details
Gene Symbol: STUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031406.2 | NP_001026577.1 | 5 - 6 | 780 | 123 | |
GenBank mRNA | AJ720999.1 | - | 5 - 6 | 123 | ||
BU208883.1 | - | 4 - 5 | 429 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
