Assay Details
Gene Symbol: TNFRSF11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033641.1 | NP_001028813.1 | 3 - 4 | 598 | 113 | |
XM_015283019.2 | XP_015138505.1 | 3 - 4 | 822 | 113 | ||
GenBank mRNA | CR352482.1 | - | 3 - 4 | 666 | 113 | |
DQ098013.1 | - | 3 - 4 | 598 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
