Assay Details
Gene Symbol: TNFRSF11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033641.1 | NP_001028813.1 | 4 - 5 | 827 | 79 | |
XM_015283019.2 | XP_015138505.1 | 4 - 5 | 1051 | 79 | ||
GenBank mRNA | CR352482.1 | - | 4 - 5 | 895 | 79 | |
DQ098013.1 | - | 4 - 5 | 827 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
