Assay Details
Gene Symbol: CETP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034814.2 | NP_001029986.2 | 5 - 6 | 579 | 68 | |
XM_025154152.1 | XP_025009920.1 | 12 - 13 | 4863 | 68 | ||
GenBank mRNA | AB205527.1 | - | 5 - 6 | 605 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
