Assay Details
Gene Symbol: CETP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034814.2 | NP_001029986.2 | 6 - 7 | 651 | 93 | |
XM_025154152.1 | XP_025009920.1 | 13 - 14 | 4935 | 93 | ||
GenBank mRNA | AB205527.1 | - | 6 - 7 | 677 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
