Assay Details
Gene Symbol: CETP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034814.2 | NP_001029986.2 | 9 - 10 | 982 | 94 | |
XM_025154152.1 | XP_025009920.1 | 16 - 17 | 5266 | 94 | ||
GenBank mRNA | AB205527.1 | - | 9 - 10 | 1008 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
