Assay Details
Gene Symbol: LIN28B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034818.1 | NP_001029990.1 | 3 - 4 | 548 | 131 | |
XM_015284525.2 | XP_015140011.1 | 4 - 5 | 726 | 131 | ||
XM_015284527.2 | XP_015140013.1 | 4 - 5 | 723 | 131 | ||
XM_015284528.2 | XP_015140014.1 | 3 - 4 | 2400 | 131 | ||
GenBank mRNA | CR353371.1 | - | 3 - 4 | 548 | 131 | |
DQ127227.1 | - | 3 - 4 | 548 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
