Assay Details
Gene Symbol: PROCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034820.1 | NP_001029992.1 | 1 - 2 | 77 | 79 | |
GenBank mRNA | AB204800.1 | - | 1 - 2 | 77 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
