Assay Details
Gene Symbol: DNASE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037838.1 | NP_001032927.1 | 5 - 6 | 750 | 93 | |
XM_015290817.2 | XP_015146303.1 | 6 - 7 | 889 | 93 | ||
GenBank mRNA | CR386219.1 | - | 5 - 6 | 750 | 93 | |
DQ272298.1 | - | 5 - 6 | 750 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
