Assay Details
Gene Symbol: CD300LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039295.1 | NP_001034384.1 | 3 - 4 | 642 | 72 | |
XM_015295393.2 | XP_015150879.1 | 3 - 4 | 944 | 72 | ||
GenBank mRNA | AM076725.1 | - | 3 - 4 | 642 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
