Assay Details
Gene Symbol: CD300LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039295.1 | NP_001034384.1 | 5 - 6 | 817 | 84 | |
XM_015295393.2 | XP_015150879.1 | 5 - 6 | 1119 | 84 | ||
GenBank mRNA | AM076725.1 | - | 5 - 6 | 817 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
