Assay Details
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039453.1 | NP_001034542.1 | 1 - 2 | 389 | 72 | |
GenBank mRNA | AM183252.1 | - | 1 - 2 | 389 | 72 | |
BX931084.1 | - | 1 - 2 | 455 | 72 | ||
BX933585.1 | - | 1 - 2 | 453 | 72 | ||
BX934345.1 | - | 1 - 2 | 454 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
