Assay Details
Gene Symbol: VAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039486.1 | NP_001034575.1 | 3 - 4 | 309 | 73 | |
XM_015293316.2 | XP_015148802.1 | 3 - 4 | 384 | 73 | ||
XM_025149125.1 | XP_025004893.1 | 3 - 4 | 384 | 73 | ||
XM_025149131.1 | XP_025004899.1 | 3 - 4 | 384 | 73 | ||
GenBank mRNA | BM490145.1 | - | 3 - 4 | 309 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
