Assay Details
Gene Symbol: GUCA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007881.2 | NP_001007882.1 | 642 | 73 | ||
GenBank mRNA | S82200.1 | - | 642 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
