Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012653.2 | NP_001012671.1 | 8 - 9 | 960 | 70 | |
GenBank mRNA | BQ038887.2 | - | 2 - 3 | 125 | 70 | |
BU204430.1 | - | 6 - 7 | 637 | 70 | ||
BU425613.1 | - | 3 - 4 | 279 | 70 | ||
BX275259.3 | - | 3 - 4 | 281 | 70 | ||
CR406506.1 | - | 8 - 9 | 969 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
