Assay Details
Gene Symbol: SELENOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024734.3 | NP_001019905.1 | 5 - 6 | 530 | 96 | |
GenBank mRNA | BX931291.2 | - | 5 - 6 | 493 | 96 | |
BX934085.2 | - | 5 - 6 | 518 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
