Assay Details
Gene Symbol: BET1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031173.2 | NP_001026344.2 | 2 - 3 | 127 | 73 | |
GenBank mRNA | AJ719590.1 | - | 2 - 3 | 127 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
