Assay Details
Gene Symbol: BRI3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040120.2 | NP_001035209.1 | 1 - 2 | 208 | 97 | |
GenBank mRNA | AB106886.1 | - | 1 - 2 | 209 | 97 | |
BX932476.2 | - | 1 - 2 | 90 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
