Assay Details
Gene Symbol: OLFM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040463.1 | NP_001035553.1 | 3 - 4 | 303 | 115 | |
GenBank mRNA | AB236174.1 | - | 3 - 4 | 303 | 115 | |
CR390719.1 | - | 2 - 3 | 167 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
