Assay Details
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044638.1 | NP_001038103.1 | 3 - 4 | 233 | 117 | |
XM_015285079.2 | XP_015140565.1 | 3 - 4 | 332 | 117 | ||
XM_025148496.1 | XP_025004264.1 | 3 - 4 | 332 | 117 | ||
GenBank mRNA | AB231849.1 | - | 3 - 4 | 233 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
