Assay Details
Gene Symbol: OPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044653.1 | NP_001038118.1 | 11 - 12 | 1667 | 117 | |
XM_015287980.1 | XP_015143466.1 | 12 - 13 | 1814 | 117 | ||
GenBank mRNA | AB255032.1 | - | 4 - 5 | 417 | 117 | |
AB295599.1 | - | 11 - 12 | 1405 | 117 | ||
AY882944.1 | - | 11 - 12 | 1667 | 117 | ||
EU124632.1 | - | 11 - 12 | 1425 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
