Assay Details
Gene Symbol: OPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044653.1 | NP_001038118.1 | 6 - 7 | 1119 | 77 | |
XM_015287980.1 | XP_015143466.1 | 7 - 8 | 1266 | 77 | ||
GenBank mRNA | AB255031.1 | - | 6 - 7 | 857 | 77 | |
AB295599.1 | - | 6 - 7 | 77 | |||
AY882944.1 | - | 6 - 7 | 1119 | 77 | ||
EU124632.1 | - | 6 - 7 | 877 | 77 | ||
EU124633.1 | - | 6 - 7 | 877 | 77 | ||
EU124634.1 | - | 6 - 7 | 877 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
