Assay Details
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048077.1 | NP_001041542.1 | 4 - 5 | 374 | 87 | |
XM_015281391.2 | XP_015136877.1 | 2 - 3 | 325 | 87 | ||
XM_015281392.2 | XP_015136878.1 | 6 - 7 | 2646 | 87 | ||
GenBank mRNA | AB243875.1 | - | 4 - 5 | 374 | 87 | |
AB243876.1 | - | 2 - 3 | 187 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
