Assay Details
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048077.1 | NP_001041542.1 | 8 - 9 | 962 | 77 | |
XM_015281391.2 | XP_015136877.1 | 6 - 7 | 913 | 77 | ||
XM_015281392.2 | XP_015136878.1 | 10 - 11 | 3234 | 77 | ||
GenBank mRNA | AB243875.1 | - | 8 - 9 | 962 | 77 | |
AB243876.1 | - | 6 - 7 | 775 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
