Assay Details
Gene Symbol: SUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079498.1 | NP_001072966.1 | 3 - 4 | 306 | 117 | |
GenBank mRNA | AJ721083.1 | - | 3 - 4 | 306 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
