Assay Details
Gene Symbol: KIFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081698.1 | NP_001075167.1 | 3 - 4 | 581 | 64 | |
GenBank mRNA | AY332739.1 | - | 3 - 4 | 581 | 64 |
Gene Symbol: KIFC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295019.2 | XP_015150505.2 | 3 - 4 | 567 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
