Assay Details
Gene Symbol: RPS3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082417.2 | NP_001075886.1 | 1 - 2 | 111 | 121 | |
GenBank mRNA | AJ737079.1 | - | 5 - 6 | 689 | 121 | |
CF250059.1 | - | 1 - 2 | 113 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
