Assay Details
Gene Symbol: RPS3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082417.2 | NP_001075886.1 | 2 - 3 | 206 | 91 | |
GenBank mRNA | AJ737079.1 | - | 4 - 5 | 594 | 91 | |
CF250059.1 | - | 2 - 3 | 208 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
