Assay Details
Gene Symbol: CALM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110364.1 | NP_001103834.1 | 5 - 6 | 476 | 147 | |
XM_025150616.1 | XP_025006384.1 | 5 - 6 | 550 | 147 | ||
GenBank mRNA | CN228694.1 | - | 5 - 6 | 147 | ||
DR430013.1 | - | 3 - 4 | 323 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
