Assay Details
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204115.2 | NP_989446.2 | 4 - 5 | 474 | 62 | |
XM_015297294.2 | XP_015152780.1 | 4 - 5 | 589 | 62 | ||
XM_015297295.2 | XP_015152781.1 | 4 - 5 | 589 | 62 | ||
GenBank mRNA | AF161712.1 | - | 4 - 5 | 442 | 62 | |
AF161713.1 | - | 4 - 5 | 474 | 62 | ||
AF507016.1 | - | 4 - 5 | 385 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
