Assay Details
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204115.2 | NP_989446.2 | 6 - 7 | 666 | 85 | |
XM_015297294.2 | XP_015152780.1 | 6 - 7 | 781 | 85 | ||
XM_015297295.2 | XP_015152781.1 | 6 - 7 | 781 | 85 | ||
GenBank mRNA | AF161712.1 | - | 6 - 7 | 634 | 85 | |
AF161713.1 | - | 6 - 7 | 666 | 85 | ||
AF507016.1 | - | 6 - 7 | 577 | 85 | ||
KU522456.1 | - | 6 - 7 | 565 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
