Assay Details
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204115.2 | NP_989446.2 | 7 - 8 | 707 | 83 | |
XM_015297294.2 | XP_015152780.1 | 7 - 8 | 822 | 83 | ||
GenBank mRNA | AF161712.1 | - | 7 - 8 | 675 | 83 | |
AF161713.1 | - | 7 - 8 | 707 | 83 | ||
AF507016.1 | - | 7 - 8 | 618 | 83 | ||
KU522456.1 | - | 7 - 8 | 606 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
