Assay Details
Gene Symbol: BRCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204276.2 | NP_989607.2 | 16 - 17 | 7778 | 67 | |
XM_025150482.1 | XP_025006250.1 | 16 - 17 | 7833 | 67 | ||
GenBank mRNA | AB066374.1 | - | 15 - 16 | 7640 | 67 | |
AY083934.1 | - | 15 - 16 | 7643 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
