Assay Details
Gene Symbol: GUCA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204320.2 | NP_989651.1 | 710 | 110 | ||
GenBank mRNA | S82199.1 | - | 710 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
