Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204400.2 | NP_989731.1 | 2 - 3 | 255 | 106 | |
GenBank mRNA | AB086192.2 | - | 2 - 3 | 303 | 106 | |
BX931986.1 | - | 2 - 3 | 356 | 106 | ||
CR354251.1 | - | 2 - 3 | 234 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
